Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012058.3 | NP_036188.1 | 2, 3 | 296 | ||
GenBank mRNA | AK011720.1 | - | 2, 3 | 224 | ||
AK020583.1 | - | 2, 3 | 245 | |||
AK020620.1 | - | 2, 3 | 237 | |||
AK051208.1 | - | 2, 3 | 242 | |||
AK146882.1 | - | 2, 3 | 245 | |||
AK169314.1 | - | 2, 3 | 244 | |||
AK187269.1 | - | 1, 2 | 71 | |||
BC039648.1 | - | 2, 3 | 283 | 5353755 | ||
BC090254.1 | - | 2, 3 | 224 | 5100716 | ||
X78304.1 | - | 2, 3 | 222 |