Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012058.3 | NP_036188.1 | 3 | 372 | ||
GenBank mRNA | AK011720.1 | - | 3 | 300 | ||
AK020583.1 | - | 3 | 321 | |||
AK020620.1 | - | 3 | 313 | |||
AK051208.1 | - | 3 | 318 | |||
AK169314.1 | - | 3 | 320 | |||
BC039648.1 | - | 3 | 359 | 5353755 | ||
BC090254.1 | - | 3 | 300 | 5100716 | ||
CJ047061.1 | - | 2 | 304 | |||
X78304.1 | - | 3 | 298 | |||
X78305.1 | - | 1 | 706 |