Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146246.3 | NP_666358.2 | 4 | 5555 | ||
GenBank mRNA | AJ491325.1 | - | 3 | 5357 | ||
AY168340.1 | - | 3 | 5336 | |||
BC031365.1 | - | 1 | 1693 | |||
BC049924.1 | - | 1 | 1693 | |||
BC129887.1 | - | 4 | 5480 | 40111348 |