Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mixl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013729.3 | NP_038757.1 | 2 | 470 | ||
GenBank mRNA | AB221547.1 | - | 2 | 452 | ||
AF135063.1 | - | 2 | 448 | |||
AF154573.1 | - | 2 | 470 | |||
AF201959.1 | - | 2 | 413 | |||
BC099876.1 | - | 2 | 470 | 40040047 | ||
BC099877.1 | - | 2 | 470 | 40040048 |