Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chaf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013733.3 | NP_038761.1 | 3 | 646 | ||
GenBank mRNA | AJ132771.2 | - | 3 | 550 | ||
AK034839.1 | - | 3 | 647 | |||
AK132551.1 | - | 470 | ||||
AK135763.1 | - | 3 | 659 | |||
BC053740.1 | - | 3 | 617 | 6504838 |