Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290830.1 | NP_001277759.1 | 27 | 5847 | ||
NM_013735.4 | NP_038763.3 | 28 | 5997 | |||
GenBank mRNA | AJ414734.1 | - | 27 | 5840 | ||
AK036324.1 | - | 9 | 1802 | |||
AK045416.1 | - | 1 | 48 | |||
AK132323.1 | - | 10 | 2030 | |||
BC035206.1 | - | 16 | 3209 | 3979746 | ||
BC079906.1 | - | 27 | 5781 | 30639375 |
Gene Symbol: Tubgcp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290824.1 | NP_001277753.1 | 18 | 3938 | ||
NM_153387.3 | NP_700436.1 | 18 | 3941 | |||
GenBank mRNA | AK046132.1 | - | 19 | 2578 | ||
AK050250.1 | - | 18 | 3939 | |||
AK152984.1 | - | 19 | 2604 | |||
AK153171.1 | - | 19 | 2604 | |||
AK161774.1 | - | 9 | 2737 |