Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173038.3 | NP_766626.1 | 4 | 514 | ||
GenBank mRNA | AK052261.1 | - | 4 | 483 | ||
AK077017.1 | - | 4 | 516 | |||
AK084854.1 | - | 4 | 527 | |||
AK212783.1 | - | 12 | ||||
BC047462.1 | - | 4 | 381 | |||
BC139386.1 | - | 4 | 512 | 8862408 | ||
BC139387.1 | - | 4 | 512 | 8862409 |