Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199004.1 | NP_001185933.1 | 12 | 2353 | ||
NM_013747.4 | NP_038775.1 | 12 | 2195 | |||
GenBank mRNA | AB016784.1 | - | 12 | 2354 | ||
AF026274.1 | - | 12 | 2185 | |||
AK138455.1 | - | 12 | 2354 | |||
AK152533.1 | - | 12 | 2203 | |||
AK153010.1 | - | 12 | 2203 | |||
AK168765.1 | - | 12 | 2199 | |||
BC016883.1 | - | 12 | 2923 | 4235952 | ||
BC086782.1 | - | 12 | 2195 | 6813878 |