Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038608.2 | NP_001033697.2 | 2 | 215 | ||
NM_001042408.1 | NP_001035867.1 | 2 | 191 | |||
NM_025299.3 | NP_079575.1 | 2 | 215 | |||
NM_178604.3 | NP_848719.2 | 2 | 327 | |||
GenBank mRNA | AK004039.1 | - | 2 | 172 | ||
AK007331.1 | - | 2 | 177 | |||
AK010599.1 | - | 2 | 215 | |||
AK011717.1 | - | 2 | 279 | |||
AK148287.1 | - | 2 | 198 | |||
AK166497.1 | - | 2 | 191 | |||
AK171121.1 | - | 2 | 167 | |||
BC003876.1 | - | 2 | 143 | |||
BC014770.1 | - | 2 | 191 | |||
BC025053.1 | - | 2 | 168 | |||
BC029327.1 | - | 2 | 185 | |||
BC031634.1 | - | 2 | 156 | 3662554 | ||
BG086914.2 | - | 1 | 104 | |||
BU514237.1 | - | 2 | 128 | |||
BY065764.1 | - | 2 | 216 | |||
BY098388.1 | - | 2 | 328 |