Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013768.3 | NP_038796.2 | 13 | 1528 | ||
GenBank mRNA | AF167573.1 | - | 13 | 1396 | ||
AK076346.1 | - | 13 | 1419 | |||
AK132414.1 | - | 13 | 1420 | |||
AK165165.1 | - | 13 | 1446 | |||
AK181096.1 | - | 2 | 138 | |||
AK187124.1 | - | 2 | 138 | |||
BC023905.1 | - | 13 | 1475 | 5321370 |