Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013768.3 | NP_038796.2 | 15 | 1779 | ||
GenBank mRNA | AF167573.1 | - | 15 | 1647 | ||
AK076346.1 | - | 15 | 1670 | |||
AK132414.1 | - | 15 | 1671 | |||
AK165165.1 | - | 15 | 1697 | |||
BC023905.1 | - | 15 | 1726 | 5321370 |