Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017404.4 | NP_059100.3 | 9 | 1028 | ||
GenBank mRNA | AK007681.1 | - | 9 | 964 | ||
AK031942.1 | - | 9 | 949 | |||
BC012274.1 | - | 9 | 907 | 3981658 |