Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177658.1 | NP_001171129.1 | 2 | 253 | ||
NM_025300.4 | NP_079576.1 | 2 | 253 | |||
GenBank mRNA | AK003026.1 | - | 2 | 234 | ||
AK003367.1 | - | 2 | 237 | |||
AK010474.1 | - | 2 | 213 | |||
AK010604.1 | - | 2 | 246 | |||
AK017820.1 | - | 2 | 232 | |||
AK017824.1 | - | 2 | 216 | |||
AK019175.1 | - | 2 | 226 | |||
AK019244.1 | - | 2 | 213 | |||
AK019249.1 | - | 2 | 213 | |||
AK150880.1 | - | 2 | 254 | |||
AK154840.1 | - | 1 | 560 | |||
AK160660.1 | - | 2 | 211 | |||
BC021304.1 | - | 2 | 249 | 3498041 | ||
BC027233.1 | - | 2 | 238 | 3498065 | ||
BC068230.1 | - | 2 | 236 | |||
BY083200.1 | - | 2 | 268 |