Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177658.1 | NP_001171129.1 | 4 | 515 | ||
NM_025300.4 | NP_079576.1 | 4 | 515 | |||
GenBank mRNA | AK003026.1 | - | 4 | 498 | ||
AK003367.1 | - | 4 | 499 | |||
AK010474.1 | - | 4 | 475 | |||
AK010604.1 | - | 4 | 508 | |||
AK011775.1 | - | 3 | 335 | |||
AK017804.1 | - | 4 | 492 | |||
AK017824.1 | - | 4 | 478 | |||
AK019175.1 | - | 4 | 488 | |||
AK019244.1 | - | 4 | 475 | |||
AK019249.1 | - | 4 | 475 | |||
AK150880.1 | - | 4 | 516 | |||
AK160660.1 | - | 4 | 473 | |||
BC021304.1 | - | 4 | 511 | 3498041 | ||
BC027233.1 | - | 4 | 500 | 3498065 | ||
BC068230.1 | - | 4 | 498 |