Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030239.2 | NP_084515.2 | 16 | 2108 | ||
GenBank mRNA | AF324242.1 | - | 16 | 2060 | ||
AK030831.1 | - | 16 | 2110 | |||
AK156626.1 | - | 16 | 2077 | |||
BC029095.1 | - | 14 | 2378 | |||
BC138772.1 | - | 16 | 2089 | 8861794 | ||
BC138773.1 | - | 16 | 2088 | 8861795 |