Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190443.1 | NP_001177372.1 | 6, 7 | 1089 | ||
NM_013853.2 | NP_038881.1 | 6, 7 | 1092 | |||
GenBank mRNA | AK087990.1 | - | 6, 7 | 994 | ||
AK137233.1 | - | 6, 7 | 1024 | |||
AK147937.1 | - | 6, 7 | 1047 | |||
AK151545.1 | - | 6, 7 | 1048 | |||
BC003300.1 | - | 5, 6 | 984 | 3583837 |