Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013788.2 | NP_038816.1 | 1 | 935 | ||
GenBank mRNA | AF148857.1 | - | 1 | 935 | ||
BC138242.1 | - | 1 | 903 | 8861261 | ||
BC138243.1 | - | 1 | 903 | 8861262 |