Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sergef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013789.2 | NP_038817.1 | 9 | 964 | ||
GenBank mRNA | AJ243952.1 | - | 9 | 947 | ||
AK145819.1 | - | 9 | 953 | |||
AK147848.1 | - | 9 | 958 | |||
BC049603.1 | - | 9 | 943 | 6591072 | ||
BC119533.1 | - | 9 | 947 | 40056360 | ||
BC119534.1 | - | 9 | 947 | 40056367 |