Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109988.1 | NP_001103458.1 | 4 | 330 | ||
NM_001109989.2 | NP_001103459.1 | 4 | 253 | |||
NM_001267717.1 | NP_001254646.1 | 4 | 242 | |||
NM_053078.4 | NP_444308.1 | 3 | 368 | |||
GenBank mRNA | AK020629.1 | - | 4 | 191 | ||
AK048361.1 | - | 3 | 168 | |||
AK081502.1 | - | 3 | 169 | |||
AK133769.1 | - | 2 | 1463 | |||
BC054762.1 | - | 3 | 150 | 6414312 | ||
BQ715224.1 | - | 3 | 155 | |||
BU057200.1 | - | 3 | 158 | |||
CJ148521.1 | - | 3 | 473 | |||
CJ165393.1 | - | 4 | 333 | |||
CX768079.1 | - | 4 | 251 | |||
EU447302.1 | - | 2 | 96 | |||
X70398.1 | - | 3 | 164 |