Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167743.1 | NP_001161215.1 | 4 | 1938 | ||
NM_181545.4 | NP_853523.2 | 6 | 2866 | |||
GenBank mRNA | AK150875.1 | - | 4 | 2188 | ||
AY261798.1 | - | 6 | 2866 | |||
BC152549.1 | - | 4 | 1938 |