Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasl10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013386.2 | NP_001013404.1 | 4 | 1171 | ||
GenBank mRNA | AK045997.1 | - | 4 | 941 | ||
AK158941.1 | - | 4 | 1172 | |||
BC116407.2 | - | 4 | 710 | 40102515 |