Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177368.4 | NP_796342.2 | 2 | 1273 | ||
GenBank mRNA | AK052344.1 | - | 2 | 1246 | ||
BC090655.1 | - | 2 | 634 | |||
BC092226.1 | - | 2 | 1229 | 6852756 | ||
BC138362.1 | - | 2 | 1004 | 8861382 | ||
BC138363.1 | - | 2 | 1004 | 8861383 |