Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030245.3 | NP_084521.1 | 7 | 1002 | ||
GenBank mRNA | AK013599.1 | - | 7 | 769 | ||
AK151132.1 | - | 7 | 864 | |||
AK180851.1 | - | 2 | 134 | |||
BC002307.1 | - | 7 | 804 | 3592046 | ||
BC027337.1 | - | 7 | 869 | 3995355 |