Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178600.2 | NP_848715.1 | 1 | 111 | ||
GenBank mRNA | AK002742.1 | - | 1 | 92 | ||
AK003237.1 | - | 1 | 112 | |||
AK008509.1 | - | 1 | 85 | |||
AK008578.1 | - | 1 | 86 | |||
AK013996.1 | - | 1 | 95 | |||
BC031732.1 | - | 1 | 93 | 3991412 |