Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178600.2 | NP_848715.1 | 1 | 176 | ||
GenBank mRNA | AK002742.1 | - | 1 | 157 | ||
AK003237.1 | - | 1 | 177 | |||
AK008509.1 | - | 1 | 150 | |||
AK008578.1 | - | 1 | 151 | |||
AK013996.1 | - | 1 | 160 | |||
BC031732.1 | - | 1 | 158 | 3991412 | ||
CD774913.1 | - | 3 | 585 |