Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kiss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178260.3 | NP_839991.2 | 2 | 157 | ||
GenBank mRNA | AA023297.1 | - | 2 | 140 | ||
AB162440.1 | - | 2 | 119 | |||
AB666166.1 | - | 3 | 317 | |||
AF472576.1 | - | 2 | 169 | |||
BC117046.1 | - | 2 | 154 | 40126365 | ||
BC119348.1 | - | 2 | 154 | 8734097 |