Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178605.4 | NP_848720.1 | 1 | 146 | ||
GenBank mRNA | AK002696.1 | - | 1 | 94 | ||
AK003769.1 | - | 1 | 128 | |||
AK013251.1 | - | 1 | 147 | |||
AK088248.1 | - | 1 | 147 | |||
BC012213.1 | - | 1 | 122 | 3964770 |