Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199009.1 | NP_001185938.1 | 10 | 1127 | ||
NM_133734.3 | NP_598495.1 | 10 | 1032 | |||
GenBank mRNA | AK033540.1 | - | 10 | 1030 | ||
AK150183.1 | - | 10 | 1119 | |||
AK152395.1 | - | 9 | 1085 | |||
AK158784.1 | - | 10 | 1112 | |||
AK170657.1 | - | 10 | 1030 | |||
BC008545.1 | - | 9 | 925 | 3497641 | ||
BC021305.1 | - | 7 | 2941 | |||
BC037001.1 | - | 9 | 1056 | 5051353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK003028.1 | - | 361 | Chr. - - [-] on Build |