Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199009.1 | NP_001185938.1 | 11 | 1333 | ||
NM_133734.3 | NP_598495.1 | 11 | 1238 | |||
GenBank mRNA | AK033540.1 | - | 11 | 1236 | ||
AK150183.1 | - | 11 | 1325 | |||
AK152395.1 | - | 10 | 1291 | |||
AK158784.1 | - | 11 | 1318 | |||
AK170657.1 | - | 11 | 1236 | |||
BC008545.1 | - | 10 | 1131 | 3497641 | ||
BC021305.1 | - | 8 | 3147 | |||
BC037001.1 | - | 10 | 1262 | 5051353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK003028.1 | - | 155 | Chr. - - [-] on Build |