Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199009.1 | NP_001185938.1 | 3 | 450 | ||
NM_133734.3 | NP_598495.1 | 3 | 355 | |||
GenBank mRNA | AK033540.1 | - | 3 | 353 | ||
AK150183.1 | - | 3 | 442 | |||
AK152395.1 | - | 2 | 408 | |||
AK158784.1 | - | 3 | 435 | |||
AK170657.1 | - | 3 | 353 | |||
AK186432.1 | - | 275 | ||||
BC008545.1 | - | 2 | 248 | 3497641 | ||
BC021305.1 | - | 1 | 1947 | |||
BC037001.1 | - | 2 | 379 | 5051353 | ||
DV648268.1 | - | 3 | 450 |