Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013865.2 | NP_038893.1 | 10 | 766 | ||
NM_180956.1 | NP_851287.1 | 10 | 766 | |||
GenBank mRNA | AB033922.1 | - | 10 | 680 | ||
AK002272.1 | - | 9 | 680 | |||
AK004689.1 | - | 10 | 712 | |||
AK036277.1 | - | 10 | 691 | |||
AK037487.1 | - | 10 | 692 | |||
AK154927.1 | - | 10 | 733 | |||
AK214669.1 | - | 6 | 336 | |||
AK218281.1 | - | 6 | 336 | |||
BC018504.1 | - | 10 | 700 | 4500970 |