Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013868.4 | NP_038896.2 | 3 | 1363 | ||
GenBank mRNA | AF155909.1 | - | 433 | |||
AJ243192.1 | - | 433 | ||||
AK030025.1 | - | 3 | 1364 | |||
AK086011.1 | - | 3 | 440 | |||
BC046993.1 | - | 3 | 399 | |||
BC089584.1 | - | 3 | 438 | 6755510 |