Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013878.2 | NP_038906.2 | 1, 2 | 106 | ||
GenBank mRNA | AF169157.1 | - | 1, 2 | 134 | ||
BC107029.2 | - | 1, 2 | 106 | 40050356 |