Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013886.4 | NP_038914.2 | 2, 3 | 594 | ||
GenBank mRNA | AB029493.1 | - | 2, 3 | 157 | ||
AK014849.1 | - | 2, 3 | 419 | |||
AK019487.1 | - | 2, 3 | 257 | |||
AK044858.1 | - | 2, 3 | 419 | |||
AK045542.1 | - | 2, 3 | 594 | |||
AK051401.1 | - | 2, 3 | 516 | |||
AK162418.1 | - | 2, 3 | 237 | |||
AK162967.1 | - | 2, 3 | 158 | |||
BC055734.1 | - | 2, 3 | 181 | 6810355 |