Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013909.2 | NP_038937.2 | 3 | 618 | ||
GenBank mRNA | AB041571.1 | - | 2 | 856 | ||
AF176522.1 | - | 3 | 606 | |||
AK040458.1 | - | 1 | 998 | |||
AK052585.1 | - | 2 | 857 | |||
AK183822.1 | - | 2 | 108 | |||
BC011485.1 | - | 1 | 991 | |||
BC026818.1 | - | 1 | 985 | |||
BC032263.1 | - | 7 | 1111 | |||
BC044784.2 | - | 2 | 818 |