Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144884.2 | NP_659133.1 | 2 | 442 | ||
GenBank mRNA | AJ298841.1 | - | 2 | 397 | ||
AK038891.1 | - | 2 | 400 | |||
AK079728.1 | - | 1 | 714 | |||
AK082936.1 | - | 2 | 352 | |||
AK160361.1 | - | 2 | 398 | |||
BC017683.1 | - | 2 | 422 | 4238480 | ||
BY724297.1 | - | 2 | 431 |