Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siva1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161737.1 | NP_001155209.1 | 3 | 430 | ||
NM_013929.2 | NP_038957.2 | 4 | 625 | |||
GenBank mRNA | AF033112.1 | - | 3 | 291 | ||
AF033113.1 | - | 4 | 486 | |||
AF033114.1 | - | 4 | 573 | |||
AK010562.1 | - | 4 | 551 | |||
AK017570.1 | - | 4 | 586 | |||
AK153115.1 | - | 4 | 535 | |||
AK169200.1 | - | 4 | 578 | |||
AK213981.1 | - | 2 | 52 | |||
AK218153.1 | - | 2 | 52 | |||
AW060433.1 | - | 1 | 169 |