Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199015.1 | NP_001185944.1 | 11 | 1225 | ||
NM_001199016.1 | NP_001185945.1 | 10 | 1113 | |||
NM_021291.3 | NP_067266.1 | 10 | 1340 | |||
GenBank mRNA | AF192310.1 | - | 11 | 1225 | ||
AJ249198.1 | - | 10 | 1340 | |||
AK142541.1 | - | 10 | 1114 | |||
AK164714.1 | - | 9 | 1530 | |||
BC010746.1 | - | 11 | 1203 | 4236578 |