Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133704.4 | NP_598465.2 | 6, 7 | 829 | ||
GenBank mRNA | AK044615.1 | - | 6, 7 | 764 | ||
AK082516.1 | - | 6, 7 | 829 | |||
BC010189.1 | - | 6, 7 | 558 | |||
BC138969.1 | - | 5, 6 | 743 | 8861991 | ||
BC138971.1 | - | 5, 6 | 743 | 8861993 | ||
BC145477.1 | - | 4, 5 | 629 |