Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist3h2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178218.4 | NP_835736.1 | 1 | 212 | ||
GenBank mRNA | AK051448.1 | - | 213 | |||
AK077568.1 | - | 1 | 213 | |||
AK082083.1 | - | 216 | ||||
AK087537.1 | - | 214 | ||||
BC058119.1 | - | 185 | 6819179 | |||
BC063781.1 | - | 179 | 6493838 |