Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175663.2 | NP_783594.1 | 1 | 92 | ||
GenBank mRNA | BC126923.1 | - | 1 | 92 | 40055824 |