Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081208.2 | NP_001074677.1 | 4 | 1846 | ||
NM_001253355.1 | NP_001240284.1 | 6 | 2010 | |||
NM_001253356.1 | NP_001240285.1 | 5 | 1951 | |||
GenBank mRNA | AK031910.1 | - | 6 | 2000 | ||
BC079867.1 | - | 4 | 1627 | |||
BC147490.1 | - | 6 | 2003 | 9056285 | ||
BC147493.1 | - | 5 | 1944 | 9056309 | ||
BC150973.1 | - | 6 | 2003 | 9087886 | ||
BC158004.1 | - | 6 | 2003 | 9088149 |