Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005510.2 | NP_001005510.2 | 98 | 17908 | ||
GenBank mRNA | AB093279.1 | - | 14 | 2025 | ||
BC042604.1 | - | 5 | 745 | |||
BC055031.1 | - | 3 | 275 | |||
BC076568.1 | - | 23 | 3482 | |||
BC082586.1 | - | 18 | 2724 |