Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpripl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033380.3 | NP_001028552.1 | 1 | 782 | ||
GenBank mRNA | AK134593.1 | - | 1 | 730 | ||
AK137678.1 | - | 1 | 784 | |||
AK149729.1 | - | 1 | 695 | |||
BC150856.1 | - | 1 | 433 | 9087767 | ||
BC150864.1 | - | 1 | 433 | 9087775 |