Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177005.4 | NP_795979.2 | 2, 3 | 236 | ||
GenBank mRNA | AK050379.1 | - | 2, 3 | 237 | ||
AK077577.1 | - | 2, 3 | 237 | |||
BY337475.1 | - | 2, 3 | 237 |