Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168684.1 | NP_001162155.1 | 3 | 1141 | ||
NM_172051.3 | NP_742048.2 | 3 | 1218 | |||
NM_177026.1 | NP_796000.1 | 3 | 1160 | |||
GenBank mRNA | AK079611.1 | - | 4 | 1078 | ||
AK083129.1 | - | 3 | 1121 | |||
AK132545.1 | - | 3 | 1196 | |||
BC057202.1 | - | 3 | 1004 | 5327878 | ||
BC095951.1 | - | 3 | 1152 | 5034018 |