Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177033.3 | NP_796007.1 | 2 | 1232 | ||
GenBank mRNA | AB292670.1 | - | 1 | 635 | ||
AK033944.1 | - | 2 | 1026 | |||
AK044771.1 | - | 2 | 767 | |||
AK141514.1 | - | 2 | 1233 | |||
BC120564.1 | - | 1 | 694 | 40129487 | ||
BC137825.1 | - | 1 | 695 | 8860844 | ||
DQ421811.1 | - | 1 | 698 |