Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A630076J17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256174.1 | NP_001243103.1 | 3 | 438 | ||
GenBank mRNA | AA119196.1 | - | 3 | 415 | ||
AI452133.1 | - | 1 | 225 | |||
AK080367.1 | - | 439 |
Gene Symbol: Prok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044344.1 | - | 2 | 2057 | ||
BC042707.1 | - | 3 | 2053 | 4975079 |