Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252094.1 | NP_001239023.1 | 5 | 899 | ||
NM_001252095.1 | NP_001239024.1 | 5 | 880 | |||
NM_001252096.1 | NP_001239025.1 | 4 | 789 | |||
NM_001252097.1 | NP_001239026.1 | 3 | 361 | |||
NM_177101.5 | NP_796075.2 | 4 | 770 | |||
GenBank mRNA | AK044252.1 | - | 2185 | |||
AK132096.1 | - | 5 | 767 | |||
BC018279.1 | - | 2 | 68 | |||
BC046911.1 | - | 3 | 300 | 6491688 | ||
BC048711.1 | - | 4 | 662 | 6328428 | ||
BC065807.1 | - | 4 | 686 | 3373106 | ||
DN174886.1 | - | 4 | 663 |