Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182808.3 | NP_877960.1 | 4, 5 | 809 | ||
GenBank mRNA | AK049880.1 | - | 3, 4 | 700 | ||
AK050001.1 | - | 4, 5 | 810 | |||
AY325120.1 | - | 4, 5 | 610 | |||
BC132179.1 | - | 4, 5 | 644 | 40130456 | ||
BC132181.1 | - | 4, 5 | 644 | 40130458 |